Learn More
MSC Tiger-Vac Pneumatic HEPA Cleaner, Mercury-Recovery, Wet/Dry, 10 gal, 1 ea
Catalog No. 502819495
Encompass
This cleaner is a HEPA, mercury-recovery, pneumatic HEPA unit suitable for both dry and wet applications. It operates on air power and features a HEPA filter within a 10-gallon polyethylene tank.
  • Operates on air power
  • HEPA filtration for contaminant capture
  • Polyethylene tank with 10 gal capacity
  • Maximum air flow of 110 CFM
  • Supports disposable filter bag, mercury recovery jar, and disposable poly liner collection
  • Includes a 1.5 in hose with a 10 ft length
  • Overall dimensions are 20 in width and 46 in height
Catalog No. 50-281-9495 Supplier MSC Supplier No. 29033891
Add to Cart
Add to Cart
Request a Quote
This cleaner is a HEPA, mercury-recovery, pneumatic HEPA unit suitable for both dry and wet applications. It operates on air power and features a HEPA filter within a 10-gallon polyethylene tank.
  • Operates on air power
  • HEPA filtration for contaminant capture
  • Polyethylene tank with 10 gal capacity
  • Maximum air flow of 110 CFM
  • Supports disposable filter bag, mercury recovery jar, and disposable poly liner collection
  • Includes a 1.5 in hose with a 10 ft length
  • Overall dimensions are 20 in width and 46 in height