Learn More
Dc Chemicals Limited MS.001 | 250mg | DC70103-250mg | Purity: >98% | 53619-67-3
Catalog No. NC2897443
Encompass_Preferred
MS.001 is a small molecule that inhibits both the chaperone binding and ubiquitin ligase activity of C-terminus of Hsc70 interacting protein (CHIP) at low micromolar concentrations
Catalog No. NC2897443 Supplier Dc Chemicals Limited Supplier No. DC70103250MG
Add to Cart
Add to Cart
MS.001 is a small molecule that inhibits both the chaperone binding and ubiquitin ligase activity of C-terminus of Hsc70 interacting protein (CHIP) at low micromolar concentrations