Learn More
Julabo USA Inc MAGIO MS-1000F 115V Refrigerated/heating circulator, -50…+200 °C, 1kW heater, 1000W cooling at 20 °C; 7.5L tank; programmable, Pt100 port, USB/RS232/RS485/Ethernet connections, optional Analog module.
SDP
Supplier:  Julabo USA Inc 9032707.02
Encompass_Preferred
MAGIO™ MS-1000F refrigerated/heating circulator with 30L tank and lid; -50…+200 °C range and 1kW heater. Cooling: 1000W @ 20 °C; 960W @ 0 °C; 510W @ -20 °C; 250W @ -30 °C; 110W @ -40 °C. For internal or external applications. Large, color touchscreen UI with built-in help functions. Cascade Control, self-optimizing for optimum results with settings for band limit, limit setting, co-speed factor and control dynamics. Variable speed pressure / suction pump 13-31 L/min; 3.5-13.3 PSI. Barbed fittings for 8 or 12mm ID tubing included (pump connections M16x1 male). Includes Pt100 connection for external monitoring/control, RS232/RS485, USB and Ethernet ports with optional slot for analog communication (requires module 9900100). Modbus TCP/IP incorporated. Stakei port for external accessory control (solenoid valve, etc). Class III (FL) according to DIN 12876-1. Integrated drain port. 115V/20A/60Hz (NEMA 5-20P plug).
Catalog No. NC1921805
Only null left
Add to Cart
Request a Quote
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov