Learn More
AdipoGen MS-1020 (1 mg)
SDP
Supplier:  AdipoGen AGCR13501M001
Encompass_Preferred
Chemical. CAS: 1255516-86-9. Formula: C21H18N2O3. MW: 346.4. Selective JAK3 inhibitor. Blocks STAT signaling. Inducer of apoptosis via down-regulation of anti-apoptotic gene expression.
Catalog No. 50-205-3876