Learn More
SAI Infusion Technologies 15 Gauge, 0.5" Blunt Needles - Non-sterile, 100/pk
SDP
Catalog No. NC0880176
Encompass
15G0.5" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 100. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.
Catalog No. NC0880176 Supplier SAI Infusion Technologies Supplier No. B1550 100 BULK
15G0.5" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 100. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.