Learn More
Instech One channel magnetic Vascular Access Button tether kit for mice. Includes: one channel mouse tether (VABM1T/25), plastic swivel (375/25PS), 24 of PU tubing, and a luer stub (LS25). Mates with one channel Vascular Access Button for mice.
SDP
Catalog No. NC1265519
Encompass
The Instech KVABM1T/25 is a one‑channel magnetic mouse VAB™ tether kit designed to mate with VABM1B/25 or VABM1B/22 buttons. It includes 24in (60cm) of PU tubing, a 25ga plastic swivel (375/25PS), 10in (25cm) PU segment, a 25ga LS25/6 luer stub, and a 7in (18cm) PS62 spring. The magnetic connector snaps into the mouse VAB™ to minimize force on the animal and support continuous infusion or sampling workflows.
Features
  • 1‑channel magnetic mouse VAB™ tether kit
  • 24in (60cm) PU tubing + 10in (25cm) PU
  • 375/25PS swivel, 25ga
  • LS25/6 luer stub, 25ga
  • 7in (18cm) PS62 spring
  • Mates with VABM1B/25 or /22
Catalog No. NC1265519 Supplier Instech Supplier No. KVABM1T/25
The Instech KVABM1T/25 is a one‑channel magnetic mouse VAB™ tether kit designed to mate with VABM1B/25 or VABM1B/22 buttons. It includes 24in (60cm) of PU tubing, a 25ga plastic swivel (375/25PS), 10in (25cm) PU segment, a 25ga LS25/6 luer stub, and a 7in (18cm) PS62 spring. The magnetic connector snaps into the mouse VAB™ to minimize force on the animal and support continuous infusion or sampling workflows.
Features
  • 1‑channel magnetic mouse VAB™ tether kit
  • 24in (60cm) PU tubing + 10in (25cm) PU
  • 375/25PS swivel, 25ga
  • LS25/6 luer stub, 25ga
  • 7in (18cm) PS62 spring
  • Mates with VABM1B/25 or /22