Learn More
Cayman Chemical MS 245oxalate 1mg
SDP
Catalog No. 502953062
Encompass_Preferred
An antagonist of 5-HT6 (Ki = 2.3 nM for the human recombinant receptor); selective for 5-HT6 over 5-HT2A and 5-HT2C receptors (Kis = 130 and 23 nM, respectively), as well as 5-HT1A, 5-HT1B, 5-HT1E, 5-HT3, and 5-HT7 receptors (Kis = 720, 9,200, 4,220, 2,390, and 600 nM, respectively); decreases cAMP production induced by 5-HT in HEK293 cells expressing 5-HT6 (pA2 = 8.88 nM); potentiates drug discrimination induced by (+)-amphetamine, but has no effect on cocaine or (–)-nicotine (Item No. 20887) drug discrimination in rats at 5 mg/kg
Catalog No. 50-295-3062 Supplier Cayman Chemical Supplier No. 119361 MG
An antagonist of 5-HT6 (Ki = 2.3 nM for the human recombinant receptor); selective for 5-HT6 over 5-HT2A and 5-HT2C receptors (Kis = 130 and 23 nM, respectively), as well as 5-HT1A, 5-HT1B, 5-HT1E, 5-HT3, and 5-HT7 receptors (Kis = 720, 9,200, 4,220, 2,390, and 600 nM, respectively); decreases cAMP production induced by 5-HT in HEK293 cells expressing 5-HT6 (pA2 = 8.88 nM); potentiates drug discrimination induced by (+)-amphetamine, but has no effect on cocaine or (–)-nicotine (Item No. 20887) drug discrimination in rats at 5 mg/kg