Learn More
AdipoGen MS-275
SDP
Catalog No. 502562386
Encompass_Preferred
Chemical. CAS 209783-80-2. Formula C21H20N4O3. MW 376.4. HDAC 1 inhibitor. Antitumor compound. Antiproliferative. TGF-beta type II receptor inducer. Apoptosis inducer. Anti-inflammatory. Angiogenesis inhibitor. Review. Promotes either self-renewal or differentiation of embryonic stem cells.
Catalog No. 50-256-2386 Supplier AdipoGen Supplier No. AGCR10032M001
Chemical. CAS 209783-80-2. Formula C21H20N4O3. MW 376.4. HDAC 1 inhibitor. Antitumor compound. Antiproliferative. TGF-beta type II receptor inducer. Apoptosis inducer. Anti-inflammatory. Angiogenesis inhibitor. Review. Promotes either self-renewal or differentiation of embryonic stem cells.