Learn More
Enzo Life Sciences MS-275 (25mg). CAS: 209783-80-2
SDP
Supplier:  Enzo Life Sciences ALX270378M025
Encompass_Preferred
Preferentially inhibits HDAC1 (IC50=300nM) over HDAC3 (IC50=8µM). Has no inhibitory activity towards HDAC8 (IC50>100µM). Alternative name: N-(2-Aminophenyl)-4-[N-(pyridine-3-ylmethoxy-carbonyl)aminomethyl]benzamide, Entinostat. Formula: C21H20N4O3. MW: 376.4. Purity: ≥95% (1H-NMR). Solubility: Soluble in DMSO or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8522