Learn More
AdipoGen MS-275 (5 mg)
SDP
Supplier:  AdipoGen AGCR10032M005
Encompass_Preferred
Chemical. CAS: 209783-80-2. Formula: C21H20N4O3. MW: 376.4. HDAC 1 inhibitor. Antitumor compound. Antiproliferative. TGF-beta type II receptor inducer. Apoptosis inducer. Anti-inflammatory. Angiogenesis inhibitor. Review. Promotes either self-renewal or differentiation of embryonic stem cells.
Catalog No. 50-205-3784