Learn More
Medchemexpress LLC MS-444 | 150045-18-4 | MFCD00923100 | 99.5% | 230.22 g/mol | C13H10O4 | 1 MG
SDP
Catalog No. 5000214423
Encompass_Preferred
MS-444 is a small-molecule inhibitor of myosin light chain kinase (MLCK) used in research to probe MLCK-dependent signaling and cytoskeletal regulation. It inhibits purified smooth muscle MLCK with an IC50 of 10 μM and is supplied as a solid or as a 10 mM solution in DMSO.
  • Inhibits MLCK with an IC50 of 10 μM.
  • High purity, approximately 99.5%.
  • Available as solid and as 10 mM solution in DMSO.
  • Molecular weight 230.22 g/mol; formula C13H10O4.
  • Stored protected from light; suitable for biochemical and cell-based research.
Catalog No. 50-002-14423 Supplier Medchemexpress LLC Supplier No. HY1006851MG
MS-444 is a small-molecule inhibitor of myosin light chain kinase (MLCK) used in research to probe MLCK-dependent signaling and cytoskeletal regulation. It inhibits purified smooth muscle MLCK with an IC50 of 10 μM and is supplied as a solid or as a 10 mM solution in DMSO.
  • Inhibits MLCK with an IC50 of 10 μM.
  • High purity, approximately 99.5%.
  • Available as solid and as 10 mM solution in DMSO.
  • Molecular weight 230.22 g/mol; formula C13H10O4.
  • Stored protected from light; suitable for biochemical and cell-based research.