Learn More
Medchemexpress LLC Ms-444 | 150045-18-4 | MFCD00923100 | 99.5% | 230.22 g/mol | C13H10O4 | 10 MG
SDP
Catalog No. 5000213749
Encompass_Preferred
MS-444 is a small-molecule inhibitor of smooth muscle myosin light chain kinase (MLCK) with reported potency around IC50 = 10 μM. Supplied for research use, it is applied in biochemical and cell-based studies to probe MLCK/myosin activity and cytoskeletal regulation. Available as a solid or as a 10 mM solution in DMSO, it has a defined molecular formula and high purity suitable for laboratory assays.
  • Inhibits MLCK with IC50 ≈ 10 μM.
  • High purity (≈99.5%).
  • Available as solid and as a 10 mM solution in DMSO.
  • Molecular formula C13H10O4; molar mass 230.22 g/mol.
  • Intended for research use in biochemical and cell-based assays.
Catalog No. 50-002-13749 Supplier Medchemexpress LLC Supplier No. HY10068510MG
MS-444 is a small-molecule inhibitor of smooth muscle myosin light chain kinase (MLCK) with reported potency around IC50 = 10 μM. Supplied for research use, it is applied in biochemical and cell-based studies to probe MLCK/myosin activity and cytoskeletal regulation. Available as a solid or as a 10 mM solution in DMSO, it has a defined molecular formula and high purity suitable for laboratory assays.
  • Inhibits MLCK with IC50 ≈ 10 μM.
  • High purity (≈99.5%).
  • Available as solid and as a 10 mM solution in DMSO.
  • Molecular formula C13H10O4; molar mass 230.22 g/mol.
  • Intended for research use in biochemical and cell-based assays.