Learn More
Julabo USA Inc MAGIO MS-600F 230V Refrigerated/heating circulator, -35…+200 °C, 2kW heater, 600W cooling at 20 °C; 7.5L tank; programmable, Pt100 port, USB/RS232/RS485/Ethernet connections, optional Analog module.
SDP
Catalog No. NC1921796
Encompass_Preferred
MAGIO™ MS-600F refrigerated/heating circulator with 7.5L tank and lid; -35…+200 °C range and 2kW heater. Cooling: 600W @ 20 °C; 440W @ 0 °C; 160W @ -20 °C. For internal or external applications. Large, color touchscreen UI with built-in help functions. Cascade Control, self-optimizing for optimum results with settings for band limit, limit setting, co-speed factor and control dynamics. Variable speed pressure / suction pump 13-31 L/min; 3.5-13.3 PSI. Barbed fittings for 8 or 12mm ID tubing included (pump connections M16x1 male). Includes Pt100 connection for external monitoring/control, RS232/RS485, USB and Ethernet ports with optional slot for analog communication (requires module 9900100). Modbus TCP/IP incorporated. Stakei port for external accessory control (solenoid valve, etc). Class III (FL) according to DIN 12876-1. Integrated drain port. 208-230V/60Hz.
Catalog No. NC1921796 Supplier Julabo USA Inc Supplier No. 9032704.33
Add to Cart
Add to Cart
Request a Quote
MAGIO™ MS-600F refrigerated/heating circulator with 7.5L tank and lid; -35…+200 °C range and 2kW heater. Cooling: 600W @ 20 °C; 440W @ 0 °C; 160W @ -20 °C. For internal or external applications. Large, color touchscreen UI with built-in help functions. Cascade Control, self-optimizing for optimum results with settings for band limit, limit setting, co-speed factor and control dynamics. Variable speed pressure / suction pump 13-31 L/min; 3.5-13.3 PSI. Barbed fittings for 8 or 12mm ID tubing included (pump connections M16x1 male). Includes Pt100 connection for external monitoring/control, RS232/RS485, USB and Ethernet ports with optional slot for analog communication (requires module 9900100). Modbus TCP/IP incorporated. Stakei port for external accessory control (solenoid valve, etc). Class III (FL) according to DIN 12876-1. Integrated drain port. 208-230V/60Hz.