Learn More
Columbia Biosciences Corp MS ANTI-6HIS DYLIGHT650 100UG
Supplier:  Columbia Biosciences Corp D101711100
Encompass_Preferred
Mouse anti-6x Histidine IgG, clone AD1.1.10, primary antibody, conjugated to DyLight 650, 100 ug. Applications include High throughput screening (HTS) and fluorescence resonance energy transfer (FRET) assays, flow cytometry
Catalog No. 50-815-66