Learn More
Enzo Life Sciences Mouse anti-biotin linker (Ready-to-use) (6ml)
SDP
Catalog No. 502012499
Encompass_Preferred
This mouse anti-biotin linker is a ready to use signal amplifying reagent optimized to increase the detection sensitivity of ENZO PATHO-GENE HPV probes. Although mouse anti-biotin linker was created to enhance ENZO probes, it will recognize any biotin associated nucleotide or macromolecule. Isotype: IgG1. Species reactivity: Species independent . Applications: ISH (in situ hybridization). Formulation: Liquid. Ready-to-use solution in TBS containing 0.05% Kathon and 1% BSA.
Catalog No. 50-201-2499 Supplier Enzo Life Sciences Supplier No. ENZ328936000
This mouse anti-biotin linker is a ready to use signal amplifying reagent optimized to increase the detection sensitivity of ENZO PATHO-GENE HPV probes. Although mouse anti-biotin linker was created to enhance ENZO probes, it will recognize any biotin associated nucleotide or macromolecule. Isotype: IgG1. Species reactivity: Species independent . Applications: ISH (in situ hybridization). Formulation: Liquid. Ready-to-use solution in TBS containing 0.05% Kathon and 1% BSA.