Learn More
Enzo Life Sciences Mouse anti-Rabbit Linker (Ready-to-use) (6ml)
SDP
Supplier:  Enzo Life Sciences ENZACC1156000
Encompass_Preferred
The Mouse anti-Rabbit Linker is a ready-to-use signal amplifying reagent optimized to increase the detection sensitivity of Enzo's POLYVIEW PLUS (anti-Mouse) Reagents. Isotype: IgG. Species reactivity: Rabbit. Applications: IHC ISH (in situ hybridization). Formulation: Ready-to-use solution in PBS containing stabilizers and preservatives.
Catalog No. 50-201-3363