Learn More
Columbia Biosciences Corp MS ANTIDYKDDDDK DYLGT488 100UG
Supplier:  Columbia Biosciences Corp D81718
Encompass_Preferred
Mouse anti-DYKDDDDK IgG, clone M2, primary antibody, conjugated to DyLight 488, 100ug. Applications include:High throughput screening (HTS) and fluorescence resonance energy transfer (FRET) assays, flow cytometry
Catalog No. 50-815-48