Learn More
Julabo USA Inc MAGIO MS-BC4 115V Heating Circulator, 4L tank, 1kW to 300 °C, programmable, Pt100 port, USB/RS232/RS485/Ethernet connections, optional Analog module. EA
SDP
Catalog No. NC1921787
Encompass_Preferred
MAGIO™ MS-BC4 heating circulator with 4L bath tank and lid with 1 kW heating power to +300 °C. Large, color touchscreen UI with built-in help functions. For internal and external temperature applications. Includes Intelligent Cascade Control, self-optimizing for optimum results with settings for band limit, limit setting, co-speed factor and control dynamics. Variable speed pressure / suction pump 13-31 L/min; 3.5-13.3 PSI. Built-in cooling coil enables counter cooling with tap water. Barbed fittings for 8 or 12mm ID tubing included (pump connections M16x1 male). Includes Pt100 connection for external monitoring/control, RS232/RS485, USB and Ethernet ports with optional slot for analog communication (requires module 9900100). Modbus TCP/IP incorporated. Stakei port for external accessory control (solenoid valve, etc). Class III (FL) according to DIN 12876-1. Includes bath lid and integrated drain port.
Catalog No. NC1921787 Supplier Julabo USA Inc Supplier No. 9032504.22
Add to Cart
Add to Cart
Request a Quote
MAGIO™ MS-BC4 heating circulator with 4L bath tank and lid with 1 kW heating power to +300 °C. Large, color touchscreen UI with built-in help functions. For internal and external temperature applications. Includes Intelligent Cascade Control, self-optimizing for optimum results with settings for band limit, limit setting, co-speed factor and control dynamics. Variable speed pressure / suction pump 13-31 L/min; 3.5-13.3 PSI. Built-in cooling coil enables counter cooling with tap water. Barbed fittings for 8 or 12mm ID tubing included (pump connections M16x1 male). Includes Pt100 connection for external monitoring/control, RS232/RS485, USB and Ethernet ports with optional slot for analog communication (requires module 9900100). Modbus TCP/IP incorporated. Stakei port for external accessory control (solenoid valve, etc). Class III (FL) according to DIN 12876-1. Includes bath lid and integrated drain port.