Learn More
Peak Scientific Instruments Ltd MS Bench SCI 2 Supports Sciex Instruments
SDP
Catalog No. 5000804181
Encompass_Preferred
Designed exclusively for SCIEX to provide modular bench solution for SCIEX Triple Quad™ 7500 LC-MS/MS System– QTRAP Ready
Noise abated compartment suitable for housing vacuum pump(s) (non generator variant only)
‘Genius Inside’: true plug & play gas generation (MS Bench (G) SCI 2 only)
No external compressed air source required
Noise & vibration dampening, suitable for use as instrumentation bench
Fan assisted cooling for vacuum pumps
Reclassifier vent preventing internal condensation (MS Bench (G) SCI 2 only)
CSA / FCC / CE compliant
Chemical-resistant phenolic resin worktop
Levelling castor wheels for seamless installation with existing lab bench system
Catalog No. 50-008-04181 Supplier Peak Scientific Instruments Ltd Supplier No. 3303000
Designed exclusively for SCIEX to provide modular bench solution for SCIEX Triple Quad™ 7500 LC-MS/MS System– QTRAP Ready
Noise abated compartment suitable for housing vacuum pump(s) (non generator variant only)
‘Genius Inside’: true plug & play gas generation (MS Bench (G) SCI 2 only)
No external compressed air source required
Noise & vibration dampening, suitable for use as instrumentation bench
Fan assisted cooling for vacuum pumps
Reclassifier vent preventing internal condensation (MS Bench (G) SCI 2 only)
CSA / FCC / CE compliant
Chemical-resistant phenolic resin worktop
Levelling castor wheels for seamless installation with existing lab bench system