Learn More
Innovative Research Inc Ms Brain Tissue Lysate
Catalog No. 502744177
Encompass_Preferred
Mouse Brain Tissue Lysate 100ug - This product is especially useful for applications like Western Blot, among others. For continuous use, store at 2-8C. If not using the entire volume immediately, store single-use aliquots at -7‘C. Repeated freeze/thaw cycles are not recommended. Under proper storage conditions, the shelf life is six months from the date of receipt. Additional Details Species Mouse Source Brain Storage Conditions -70C
Catalog No. 50-274-4177 Supplier Innovative Research Inc Supplier No. IMSBRTL100UG
Mouse Brain Tissue Lysate 100ug - This product is especially useful for applications like Western Blot, among others. For continuous use, store at 2-8C. If not using the entire volume immediately, store single-use aliquots at -7‘C. Repeated freeze/thaw cycles are not recommended. Under proper storage conditions, the shelf life is six months from the date of receipt. Additional Details Species Mouse Source Brain Storage Conditions -70C