Learn More
Waters Corp MS Calibration Reference Standard - PEG 400
Catalog No. 50201686 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Mass Spectrometry standard for system assessment. 1 X 10 mL Calibration Solution PEG 400: PEG 400 10ug/mL, Ammonium Acetate 30.8ug/mL in Acetonitrile/Water 1:1 v/v, 15mL Cear Nalgene Bottle Certificate of Analysis
Catalog No. 50-201-686 Supplier Waters Corp Supplier No. 700004595
Mass Spectrometry standard for system assessment. 1 X 10 mL Calibration Solution PEG 400: PEG 400 10ug/mL, Ammonium Acetate 30.8ug/mL in Acetonitrile/Water 1:1 v/v, 15mL Cear Nalgene Bottle Certificate of Analysis