Learn More
Waters Corp MS Qualification Standards Kit
Supplier:  Waters Corp 700002157
Encompass_Preferred
Standards Kit used for Mass Spec. The kit has 5 Standards including1. 2 of 10mL Solution of 250ug/L Daidzein in 30/70 AcetoN/Water in 15mL clear nalgene2. 15mL of 2ng/ul Reserpine, 25ng/ul PPG 1000 50ng/ul Triacetyl-B-Cyclodextrin in 50/50 Aceto/4mM Ammonium Acetate in clear 15mL Nalgene Bottle3. 15mL of 2ug/ul, +1%, -0%Sodium Iodide, 50ng/ul, Cesium Iodide on 50/50 IPA Water in clear 15mL Nalgene4. 10ml Solution, 1 mg/L Daidzein in 30/70 Aceto/Water in clear 15mL Nalgene5. 10ml Solution, Formic Acid in 15mL Brown Nalgene
Catalog No. 50-792-695