Learn More
Innovative Research Inc Ms Comp Fctr B Kit
Supplier:  Innovative Research Inc IMSCFBKT
Encompass_Preferred
Mouse Complement Factor B ELISA Kit - Mouse Complement Factor B ELISA Kit from Innovative Research is intended for the quantitative determination of Mouse Complement Factor B in biofluid samples, such as serum, plasma and other biological fluids. This is a sandwich ELISA in strip well format with reagents for up to 96 tests . CFB present in samples reacts with an anti-Mouse CFB capture antibody coated and dried on a microtiter plate. After appropriate washing steps, an anti-CFB detection antibody conjugated to biotin binds to the captured analyte. Excess antibody is washed away and bound antibody is then reacted with avidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. Color development is directly proportional to the concentration of active CFB in the samples. A standard calibration curve is prepared along with the samples to be measured using dilutions of Mouse CFB.
Catalog No. 50-274-4171