Learn More
Innovative Research Inc Ms Creatine Kinase Muscle Kit
Catalog No. 502744170
Encompass_Preferred
Mouse Creatine Kinase Muscle ELISA Kit - Mouse Creatine Kinase Muscle ELISA Kit from Innovative Research is intended for the quantitative determination of Mouse Creatine Kinase, Muscle in biofluid samples, such as serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids. This is a sandwich ELISA in strip well format with reagents for up to 96 tests . CKM present in samples reacts with an anti-Mouse CKM capture antibody coated and dried on a microtiter plate. After appropriate washing steps, an anti-CKM detection antibody conjugated to biotin binds to the captured analyte. Excess antibody is washed away and bound antibody is then reacted with avidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. Color development is directly proportional to the concentration of active CKM in the samples. A standard calibration curve is prepared along with the samples to be measured using dilutions of Mouse CKM.
Catalog No. 50-274-4170 Supplier Innovative Research Inc Supplier No. IMSCKMKT
Mouse Creatine Kinase Muscle ELISA Kit - Mouse Creatine Kinase Muscle ELISA Kit from Innovative Research is intended for the quantitative determination of Mouse Creatine Kinase, Muscle in biofluid samples, such as serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids. This is a sandwich ELISA in strip well format with reagents for up to 96 tests . CKM present in samples reacts with an anti-Mouse CKM capture antibody coated and dried on a microtiter plate. After appropriate washing steps, an anti-CKM detection antibody conjugated to biotin binds to the captured analyte. Excess antibody is washed away and bound antibody is then reacted with avidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. Color development is directly proportional to the concentration of active CKM in the samples. A standard calibration curve is prepared along with the samples to be measured using dilutions of Mouse CKM.