Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters MS ETD Reagent Kit, MS Detection, Solid, 12 Month Shelf Life

Catalog No. 50201697 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The MS ETD Reagent Kit is a Mass Spectrometry standard designed for system assessment. It is supplied as a solid formulation and includes 5 X Nitrosobenzene 100 mg ± 5 mg in 4 mL Amber Screw top Vials, ensuring reliable performance for various applications.

  • Detection method: MS
  • Formulation: Solid
  • Storage condition: Room temperature
  • System type: MS
  • Instrument: Synapt GS
  • Shelf life: 12 months
  • Shelf life upon reconstitution: 1 week
  • Shipping condition: Ambient
  • Tolerance: 0.05
  • Mass: 100 mg
  • Type: Reference standard
  • Application: System performance
  • Product type: Standard kits
  • Units per package: 1 pk
  • Contents: 5 X Nitrosobenzene 100 mg ± 5 mg 4 mL Amber Screw top Vial
  • Hazardous material: This product contains hazardous material which requires special freight handling; additional charges may apply.

Catalog No. 50-201-697 Supplier Waters Corp Supplier No. 700005669
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MS ETD Reagent Kit is a Mass Spectrometry standard designed for system assessment. It is supplied as a solid formulation and includes 5 X Nitrosobenzene 100 mg ± 5 mg in 4 mL Amber Screw top Vials, ensuring reliable performance for various applications.

  • Detection method: MS
  • Formulation: Solid
  • Storage condition: Room temperature
  • System type: MS
  • Instrument: Synapt GS
  • Shelf life: 12 months
  • Shelf life upon reconstitution: 1 week
  • Shipping condition: Ambient
  • Tolerance: 0.05
  • Mass: 100 mg
  • Type: Reference standard
  • Application: System performance
  • Product type: Standard kits
  • Units per package: 1 pk
  • Contents: 5 X Nitrosobenzene 100 mg ± 5 mg 4 mL Amber Screw top Vial
  • Hazardous material: This product contains hazardous material which requires special freight handling; additional charges may apply.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.