Learn More
Innovative Research Inc Ms FXI Hvy Chn moAbs 12B11
Catalog No. 502744207
Encompass_Preferred
Mouse Anti Human Factor XI Heavy Chain Monoclonal Clone 12B11 100ug - Mouse Anti Human Factor XI Heavy Chain Monoclonal Clone 12B11 from Innovative Research is a monoclonal antibody in a Frozen liquid format, buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. This antibody has been purified using protein G affinity chromatography. This product can be used in analytic tools such as ELISA kits and Western blot. This product detects Factor XI and Factor XIa under non-reducing and reducing conditions. The epitope is localized to the heavy chain. Clone 2B11-E5F6, isotype IgG1 Kappa.
Catalog No. 50-274-4207 Supplier Innovative Research Inc Supplier No. IMSAHUFXIHC12B11C100UG
Mouse Anti Human Factor XI Heavy Chain Monoclonal Clone 12B11 100ug - Mouse Anti Human Factor XI Heavy Chain Monoclonal Clone 12B11 from Innovative Research is a monoclonal antibody in a Frozen liquid format, buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. This antibody has been purified using protein G affinity chromatography. This product can be used in analytic tools such as ELISA kits and Western blot. This product detects Factor XI and Factor XIa under non-reducing and reducing conditions. The epitope is localized to the heavy chain. Clone 2B11-E5F6, isotype IgG1 Kappa.