Learn More
Innovative Research Inc Ms FXII Hvy Chn moAbs 11B9
Catalog No. 502744206
Encompass_Preferred
Mouse Anti Human Factor XII Heavy Chain Monoclonal Clone 11B9 100ug - Mouse Anti Human Factor XII Heavy Chain Monoclonal Clone 11B9 from Innovative Research is a monoclonal antibody in a Frozen liquid format, buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. This antibody has been purified using protein G affinity chromatography. This product can be used in analytic tools such as ELISA kits and Western blot. This product detects Factor XII, Factor XIIa and Factor beta-XIIa under non-reducing conditions. The epitope is localized to the heavy chain between residues Ile 1 and Thr 279. Clone 11B9-4F8, isotype IgG1 Kappa.
Catalog No. 50-274-4206 Supplier Innovative Research Inc Supplier No. IMSAHUFXIIHC11B9C100UG
Mouse Anti Human Factor XII Heavy Chain Monoclonal Clone 11B9 100ug - Mouse Anti Human Factor XII Heavy Chain Monoclonal Clone 11B9 from Innovative Research is a monoclonal antibody in a Frozen liquid format, buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. This antibody has been purified using protein G affinity chromatography. This product can be used in analytic tools such as ELISA kits and Western blot. This product detects Factor XII, Factor XIIa and Factor beta-XIIa under non-reducing conditions. The epitope is localized to the heavy chain between residues Ile 1 and Thr 279. Clone 11B9-4F8, isotype IgG1 Kappa.