Learn More
Innovative Research Inc Ms FXII Hvy Chn moAbs 20C8
Catalog No. 502744204
Encompass_Preferred
Mouse Anti Human Factor XII Heavy Chain Monoclonal Clone 20C8 100ug - Mouse Anti Human Factor XII Heavy Chain Monoclonal Clone 20C8 from Innovative Research is a monoclonal antibody in a Frozen liquid format, buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. This antibody has been purified using protein G affinity chromatography. This product can be used in analytic tools such as ELISA kits and Western blot. This product detects Factor XII and Factor XIIa under non-reducing and reducing conditions. This epitope is localized to the heavy chain. Clone 20C8-6F9F1, isotype IgG1 Kappa.
Catalog No. 50-274-4204 Supplier Innovative Research Inc Supplier No. IMSAHUFXIIHC20C8C100UG
Mouse Anti Human Factor XII Heavy Chain Monoclonal Clone 20C8 100ug - Mouse Anti Human Factor XII Heavy Chain Monoclonal Clone 20C8 from Innovative Research is a monoclonal antibody in a Frozen liquid format, buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. This antibody has been purified using protein G affinity chromatography. This product can be used in analytic tools such as ELISA kits and Western blot. This product detects Factor XII and Factor XIIa under non-reducing and reducing conditions. This epitope is localized to the heavy chain. Clone 20C8-6F9F1, isotype IgG1 Kappa.