Learn More
Enzo Life Sciences MULTIVIEW PLUS (mouse-HRP/rabbit-AP) IHC Kit (Brown/Green) (150 tests)
SDP
Supplier:  Enzo Life Sciences ENZKIT1810150
Encompass_Preferred
The MULTIVIEW PLUS (mouse-HRP/rabbit-AP) IHC Kit is a non-biotin one-step detection system suitable for detecting and localizing antigens in formalin-fixed paraffin-embedded tissues. Assay Time: ~2 hours. Applications: IHC .
Catalog No. 50-201-3859