Learn More
Innovative Research Inc Ms IgG Depleted Srm 100ml
Catalog No. 502744152
Encompass_Preferred
Mouse IgG Depleted Serum 100ml - Mouse IgG Depleted Serum from Innovative Research is purified from normal mouse serum using affinity chromatography. This product can be used as a positive control for ELISA and western blot and for a variety of in vitro applications including tissue culture and more. Prepared from frozen US origin CD-1 mouse serum using immobilized Protein A.
Catalog No. 50-274-4152 Supplier Innovative Research Inc Supplier No. IMSIGGDS100ML
Mouse IgG Depleted Serum 100ml - Mouse IgG Depleted Serum from Innovative Research is purified from normal mouse serum using affinity chromatography. This product can be used as a positive control for ELISA and western blot and for a variety of in vitro applications including tissue culture and more. Prepared from frozen US origin CD-1 mouse serum using immobilized Protein A.