Learn More
AdipoGen Mouse IgG1 Isotype Control (Biotin) (50 µg)
SDP
Supplier:  AdipoGen AG35B0003BC050
Encompass_Preferred
Monoclonal Antibody. Liquid. In PBS containing 0.02% sodium azide. Applications: FACS, ICC, IHC, IP. Negative control in assays where mouse IgG1 antibodies labeled to Biotin are used.
Catalog No. 50-205-3054