Learn More
AdipoGen Mouse IgG1 Isotype Control (preservative free) (500 µg)
SDP
Supplier:  AdipoGen AG35B0003PFC500
Encompass_Preferred
Monoclonal Antibody. Isotype: Mouse IgG1. Clone: 15H6. Applications: FACS, ICC, IHC, IP. Liquid. In PBS. Negative control in assays where mouse IgG1 antibodies are used.
Catalog No. 50-205-3055