Learn More
AdipoGen ms IgG1 Isotype Ctrl
SDP
Supplier:  AdipoGen AG35B0003PFC050
Encompass_Preferred
Monoclonal Antibody. Isotype Mouse IgG1. Clone 15H6. Applications FACS, ICC, IHC, IP. Liquid. In PBS. Negative control in assays where mouse IgG1 antibodies are used.
Catalog No. 50-256-3093