Learn More
AdipoGen ms IgG1 Isotype Ctrl A647N
SDP
Supplier:  AdipoGen AG35B0003TSC050
Encompass_Preferred
Monoclonal Antibody. Isotype Mouse IgG1. Clone 15H6. Applications FACS, ICC. Liquid. In PBS containing 0.02% sodium azide. Negative control in assays where mouse IgG1 ATTO 647N-labeled antibodies are used.
Catalog No. 50-256-3091