Learn More
Enzo Life Sciences Mouse IgG2a kappa Isotype control, Recombinant monoclonal antibody (MOPC-21) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5770200
Encompass_Preferred
Clone: MOPC-21. Isotype: IgG2aκ. Applications: Flow Cytometry, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3103