Learn More
Bon Opus Biosciences Recombinant Mouse IL-11 (N-hFc) - Source: Human 293 Cells
SDP
Supplier:  Bon Opus Biosciences BP25950UG
Encompass_Preferred
Recombinant Mouse IL-11 (N-Fc) is produced by our Mammalian expression system and the target gene encoding Pro22-Leu199 is expressed with a Fc tag at the N-terminus. Accession#P47873. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4
Catalog No. NC2733742