Learn More
Innovative Research Inc Ms KNG Lt Chn moAbs 17E10
Catalog No. 502744202
Encompass_Preferred
Mouse Anti Human Kininogen Light Chain Monoclonal Clone 17E10 100ug - Mouse Anti Human Kininogen Light Chain Monoclonal Clone 17E10 from Innovative Research is a monoclonal antibody in a Frozen liquid format, buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. This antibody has been purified using protein G affinity chromatography. This product can be used in analytic tools such as ELISA kits and Western blot. This product strongly blots kininogen under non-reducing and reducing conditions. The binding epitope is on the light chain of kininogen. Clone 17E10-G9, isotype IgG2b.
Catalog No. 50-274-4202 Supplier Innovative Research Inc Supplier No. IMSAHUKNGLC17E10C100UG
Mouse Anti Human Kininogen Light Chain Monoclonal Clone 17E10 100ug - Mouse Anti Human Kininogen Light Chain Monoclonal Clone 17E10 from Innovative Research is a monoclonal antibody in a Frozen liquid format, buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. This antibody has been purified using protein G affinity chromatography. This product can be used in analytic tools such as ELISA kits and Western blot. This product strongly blots kininogen under non-reducing and reducing conditions. The binding epitope is on the light chain of kininogen. Clone 17E10-G9, isotype IgG2b.