Learn More
Biocare Medical, LLC MS-ON-CANINE HRP-POLYMER 25ML
SDP
Catalog No. 5082920
Encompass_Preferred
Mouse-on-Canine HRP-Polymer is designed as a one-step detection for mouse antibodies on canine and feline tissues. Combine together for universal or simultaneous double stains. It is labeled with horseradish peroxidase (HRP).
Catalog No. 50-829-20 Supplier Biocare Medical, LLC Supplier No. MC541H
Mouse-on-Canine HRP-Polymer is designed as a one-step detection for mouse antibodies on canine and feline tissues. Combine together for universal or simultaneous double stains. It is labeled with horseradish peroxidase (HRP).