Learn More
Medchemexpress LLC Ms-PEG4-MS | 55400-73-2 | 95.0% | 350.41 | 500 MG
SDP
Catalog No. 5000404401
Encompass_Preferred
Ms-PEG4-MS is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are molecules that contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other is for the target protein. They function by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-004-04401 Supplier Medchemexpress LLC Supplier No. HY42774500MG
Ms-PEG4-MS is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are molecules that contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other is for the target protein. They function by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only