Learn More
Medchemexpress LLC Ms-PEG5-t-butyl ester | 870487-48-2 | C₁₆H₃₂O₉S | 250 MG
SDP
Catalog No. 5000352584
Encompass_Preferred
Ms-PEG5-t-butyl ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-52584 Supplier Medchemexpress LLC Supplier No. 250MGHY140384
Ms-PEG5-t-butyl ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.