Learn More
Medchemexpress LLC Ms-PEG5-t-butyl ester | 870487-48-2 | C16H32O9S | 50 MG
SDP
Catalog No. 5000356987
Encompass_Preferred
Ms-PEG5-t-butyl ester is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase, and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system.
  • Used to selectively degrade target proteins.
Catalog No. 50-003-56987 Supplier Medchemexpress LLC Supplier No. HY14038450MG
Ms-PEG5-t-butyl ester is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase, and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system.
  • Used to selectively degrade target proteins.