Learn More
Echelon Biosciences Research Labs MS-PTDINS(4 5)P2 DIC16 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs PF4516100UG
Encompass
PtdIns(4,5)P2 a-fluorophosphonate diC16 (ms-PtdIns(4,5)P2 di16C) is a synthetic PI(4,5)P2 analog in which the phosphodiester bond has been rendered metabolically stable via the -fluorophosphonate. P-F4508 restores the TRPM4 ion channel current at 10M. This item is non returnable
Catalog No. 50-720-700