Learn More
Biocare Medical, LLC MS&RB-ON-RODENT POLYMER 25ML
SDP
Catalog No. 5083273
Encompass_Preferred
Mouse-&-Rabbit-on-Rodent Double Stain Polymer is a simultaneous detection is for a mouse and rabbit antibody cocktail on mouse or rat tissue with HRP and AP labels. The double staining procedure can be completed in approximately 2 hours.
Catalog No. 50-832-73 Supplier Biocare Medical, LLC Supplier No. RDS513H
Mouse-&-Rabbit-on-Rodent Double Stain Polymer is a simultaneous detection is for a mouse and rabbit antibody cocktail on mouse or rat tissue with HRP and AP labels. The double staining procedure can be completed in approximately 2 hours.