Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp MS Reference Standards, APGC/GC-MS, Liquid, Solid

Catalog No. 502392978 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Waters Corporation MS Reference Standards are designed for Atmospheric Pressure GC-MS applications, ideal for system validation and calibration. This product includes two each of: 1 mL 1mg/mL HCB in Hexane, 1 mL heptacosafluorotributylamine, 1 mL 1000 pg/μL phenanthrene in hexane, and 1 mL 1000 pg/μL 4-bromophenylphenylether in hexane. Compatible with GCMS systems and APGC instruments.

  • Separation mode: GC.
  • Detection method: MS.
  • Formulation: Liquid, solid.
  • Storage: 4°C (Fridge).
  • System type: GCMS.
  • Instrument: APGC.
  • Shelf life: 12 months.
  • Shipping: Ambient.
  • Type: Reference.
  • Applications: System performance, calibration.
  • Product type: Standards.
  • Units per package: 1 pk.
  • Contains hazardous material; special freight and charges apply.

Catalog No. 50-239-2978 Supplier Waters Corp Supplier No. 700005254
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Waters Corporation MS Reference Standards are designed for Atmospheric Pressure GC-MS applications, ideal for system validation and calibration. This product includes two each of: 1 mL 1mg/mL HCB in Hexane, 1 mL heptacosafluorotributylamine, 1 mL 1000 pg/μL phenanthrene in hexane, and 1 mL 1000 pg/μL 4-bromophenylphenylether in hexane. Compatible with GCMS systems and APGC instruments.

  • Separation mode: GC.
  • Detection method: MS.
  • Formulation: Liquid, solid.
  • Storage: 4°C (Fridge).
  • System type: GCMS.
  • Instrument: APGC.
  • Shelf life: 12 months.
  • Shipping: Ambient.
  • Type: Reference.
  • Applications: System performance, calibration.
  • Product type: Standards.
  • Units per package: 1 pk.
  • Contains hazardous material; special freight and charges apply.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.