Learn More
Waters Corp MS Reference Standard/ Reverse Peptide Kit
Catalog No. 50201689 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The HDMS Standard Kit - Reversed Peptide Kit contains three well characterized samples intended to calibrate ion mobility spectrometry/mass spectrometry (IMS/MS) systems. Additionally, this standard can be utilized as a reference standard for verifying IMS/MS system performance.
The kit contains the following three components:
1 vial Ser-Asp-Gly-Arg-Gly, 1 mg in a 4 mL Amber Glass Screw Top Vial
1 vial Gly-Arg-Gly-Asp-Ser, 1 mg in a 4 mL Amber Glass Screw Top Vial
1 vial Poly-DL-Alanine, 1 mg in a 4 mL Amber Glass Screw Top Vial
Store at -20ºC ± 10°C (Frozen)
Catalog No. 50-201-689 Supplier Waters Corp Supplier No. 700005089
The HDMS Standard Kit - Reversed Peptide Kit contains three well characterized samples intended to calibrate ion mobility spectrometry/mass spectrometry (IMS/MS) systems. Additionally, this standard can be utilized as a reference standard for verifying IMS/MS system performance.
The kit contains the following three components:
1 vial Ser-Asp-Gly-Arg-Gly, 1 mg in a 4 mL Amber Glass Screw Top Vial
1 vial Gly-Arg-Gly-Asp-Ser, 1 mg in a 4 mL Amber Glass Screw Top Vial
1 vial Poly-DL-Alanine, 1 mg in a 4 mL Amber Glass Screw Top Vial
Store at -20ºC ± 10°C (Frozen)