Learn More
Laxco™ MS Series Fixed Magnification Stereo Microscope System
SDP
Catalog No. MS20415 Shop All Laxco Inc Products
Change view
Click to view available options
Magnification Power:
10X to 30X
15X to 45X
20X to 40X
40X to 80X
Eyepiece:
10X
15X
20X
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power Eyepiece
MS20415 15X to 45X 15X
MS20420 40X to 80X 20X
MS204 20X to 40X 10X
MS103 10X to 30X 10X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS20415 Supplier Laxco™ Supplier No. MS20415
Utilizes fixed dual magnification optical system. Laxco™ MS Series Fixed Magnification Stereo Microscope System are research-grade instrument with wide choice of zoom magnification ranges and most commonly found in general laboratory.
Optics
  • Choose between multiple combinations of magnifications
  • Dual diopter adjustments accommodates different eye focal length
  • Parfocality maintained between magnification
  • Clearly marked neural knobs for magnification
Stand
  • Rugged, all metal construction
  • Large, contoured stage
  • Left and right hand operation
  • Slip clutch prevents damage to the focusing mechanism
  • Locked-in stage clip
Light Source
  • 20w halogen lower illumination
  • 15w halogen, with reflector for upper illumination
  • Individually control light sources
Eyepiece
  • 10X super-widefield eyepieces with interpupillary and diopter adjustments to correct for user-specific eyesight
Certifications
Quality Proformance Certification
Specifications
Magnification Power 15X to 45X
Warranty 2 Year - Electronic, Mechanical, Optical with option to extend to 5 years
Product Type Stereo Microscope
Lighting Type Transmitted Fluorescent and Halogen Reflected
Eyepiece 15X