Learn More
Waters Corp Waters Corporation MS Set-up Solution, Atmospheric Pressure Ionization, Reversed Phase, 15 mL
Catalog No. 50201659 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This standard solution is used for mass spectrometer set-up, containing 2ng/μL Reserpine, 25ng/μL PPG 1000, and 50ng/μL Triacetyl-B-Cyclodextrin in a 50/50 Acetonitrile/Water mixture with 4mM Ammonium Acetate. It is supplied in a 15mL Nalgene bottle and supports accurate sample preparation in liquid chromatography.
  • Standard solution for mass spectrometry set-up, including system performance and calibration.
  • Composed of 2ng/μL Reserpine, 25ng/μL PPG 1000, and 50ng/μL Triacetyl-B-Cyclodextrin.
  • Formulated in 50/50 Acetonitrile/Water with 4mM Ammonium Acetate.
  • Provided in a 15mL Nalgene bottle.
  • Features reversed phase separation and MS detection.
  • Stable for 18 months with room temperature storage and ambient shipping.
  • Contains hazardous material, potentially incurring special freight handling and additional charges.
Catalog No. 50-201-659 Supplier Waters Corp Supplier No. 700001594
This standard solution is used for mass spectrometer set-up, containing 2ng/μL Reserpine, 25ng/μL PPG 1000, and 50ng/μL Triacetyl-B-Cyclodextrin in a 50/50 Acetonitrile/Water mixture with 4mM Ammonium Acetate. It is supplied in a 15mL Nalgene bottle and supports accurate sample preparation in liquid chromatography.
  • Standard solution for mass spectrometry set-up, including system performance and calibration.
  • Composed of 2ng/μL Reserpine, 25ng/μL PPG 1000, and 50ng/μL Triacetyl-B-Cyclodextrin.
  • Formulated in 50/50 Acetonitrile/Water with 4mM Ammonium Acetate.
  • Provided in a 15mL Nalgene bottle.
  • Features reversed phase separation and MS detection.
  • Stable for 18 months with room temperature storage and ambient shipping.
  • Contains hazardous material, potentially incurring special freight handling and additional charges.