Learn More
BD Ms Single Cell Sample Multiplexing Kit
Catalog No. BDB940517 Shop All BD Biosciences Products
Change view
Click to view available options
Product Type:
Ms Single Cell Sample Multiplexing Kit
Catalog No. Product Type
BDB940517 Ms Single Cell Sample Multiplexing Kit
1 options
Catalog No. BDB940517 Supplier BD Supplier No. 633793
Add to Cart
Add to Cart
Provides the necessary reagents to support labelling Sample Tags on mouse immune single cells.
The BD™ Ms Single-Cell Multiplexing Kit allows you to combine and simultaneously process up to 12 different samples in one single-cell multiomics experiment on the BD Rhapsody™Single-Cell Analysis System. The kits are designed to work with all BD Rhapsody™ Assays and include streamlined informatics tools integrated into the BD Rhapsody™ Bioinformatics Analysis Pipeline to automatically demultiplex by Sample Tags and identify individual samples during data analyses.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Ms Single Cell Sample Multiplexing Kit