Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters MS Solution, Daidzein (1 mg/L), 10 mL, Reference

Catalog No. 50201663 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This MS solution contains Daidzein at 1 mg/L, prepared in a 30/70 Acetonitrile Water mixture within a 15 mL clear Nalgene container. It is designed for use as a standard in Mass Spectrometry applications.

  • Used for Mass Spec applications
  • Contains 1 mg/L Daidzein
  • Formulated as a liquid
  • Volume: 10 mL
  • Separation mode: Reversed Phase
  • Detection method: MS
  • Storage condition: Room Temperature
  • System type: MS
  • Shelf life: 18 Months
  • Shipping condition: Ambient
  • Type: Reference
  • Application: System Performance
  • Product type: Standards
  • Units per package: 1 pk
  • This product contains hazardous material which requires special freight handling. Additional charges may apply.

Catalog No. 50-201-663 Supplier Waters Corp Supplier No. 700002340
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This MS solution contains Daidzein at 1 mg/L, prepared in a 30/70 Acetonitrile Water mixture within a 15 mL clear Nalgene container. It is designed for use as a standard in Mass Spectrometry applications.

  • Used for Mass Spec applications
  • Contains 1 mg/L Daidzein
  • Formulated as a liquid
  • Volume: 10 mL
  • Separation mode: Reversed Phase
  • Detection method: MS
  • Storage condition: Room Temperature
  • System type: MS
  • Shelf life: 18 Months
  • Shipping condition: Ambient
  • Type: Reference
  • Application: System Performance
  • Product type: Standards
  • Units per package: 1 pk
  • This product contains hazardous material which requires special freight handling. Additional charges may apply.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.