Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters MS Solution, Daidzein, 250 μg/L, Liquid

Catalog No. 50201662 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This product consists of standards used for Mass Spec. It contains 2 x 10 mL of 250 μg/L Daidzein in a 30/70 Acetonitrile/Water mixture, packaged in a 15 mL clear nalgene bottle.

  • Separation mode: Reversed phase
  • Detection method: MS
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: MS
  • Shelf life: 18 months
  • Shipping condition: Ambient
  • Volume: 10 mL
  • Type: Reference
  • Application: System performance
  • Product type: Standards
  • Units per package: 1 pk
  • Contains hazardous material which requires special freight handling; additional charges may apply

Catalog No. 50-201-662 Supplier Waters Corp Supplier No. 700002339
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product consists of standards used for Mass Spec. It contains 2 x 10 mL of 250 μg/L Daidzein in a 30/70 Acetonitrile/Water mixture, packaged in a 15 mL clear nalgene bottle.

  • Separation mode: Reversed phase
  • Detection method: MS
  • Formulation: Liquid
  • Storage condition: Room temperature
  • System type: MS
  • Shelf life: 18 months
  • Shipping condition: Ambient
  • Volume: 10 mL
  • Type: Reference
  • Application: System performance
  • Product type: Standards
  • Units per package: 1 pk
  • Contains hazardous material which requires special freight handling; additional charges may apply

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.